Learn More
Hach Company Stablcal Turbidity Standards Calibration Set, 2100P Portable Turbidimeter, 100 mL bottles
Supplier: Hach Company 2659410
Stablcal Calibration Standards Kit, for 2100P Portable Turbidimeter. USEPA accepted for calibration purposes. Stablcal Stabilized Formazin Turbidity Standards are true Formazin dilutions developed for use in any turbidimeter. Kit includes 100 mL bottles of 0.1, 20, 100 and 800 NTU standard.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.