Learn More
Hach Company Turbidity Standard, Stablcal, 400.0 NTU, 500 mL
Supplier:  Hach Company 7121649
Encompass_Preferred
Stablcal Turbidity Standard Bulk Solution Calibration Kits for Hach 1720 Series Turbidimeters. USEPA accepted for calibration purposes Stablcal Stabilized Formazin Turbidity Standards are true Formazin dilutions developed for use in any turbidimeter.
Catalog No. 50-199-7222
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.