Learn More
Bellco Glass, Inc. Sst Weight Platform 12X12"X3" 3 L Bio
SDP
Supplier:  Bellco Glass, Inc. 780512125
Encompass_Preferred
Weight platforms allow the user to monitor and/or control the weight of the reaction vessels. The platforms contain 4 load cells and plugs into the rear of the CellTrol II units having weight capability.
Catalog No. 50-194-6361
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.