Learn More
Bellco Glass, Inc. Sst Pipet Can W/ Sil Pad 5X5X41Cm
SDP
Supplier:  Bellco Glass, Inc. 121711116
Encompass_Preferred
Heavy gauge rectangular construction for easy stacking and storage. These pipette cans can be sterilized by autoclave or dry heat. (do not exceed 400 degrees f). Includes silicone pad on the inside of top and bottom to protect pipette tips.
Catalog No. 50-194-4603
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.