Learn More
New England Biolabs, Inc. ssRNA Ladder – 25 gel lanes
Supplier: New England Biolabs, Inc. N0362S
The ssRNA Ladder is a set of 7 RNA molecules produced by in vitro transcription of a mixture of 7 linear DNA templates. The ladder sizes are: 9000, 7000, 5000, 3000, 2000, 1000 and 500 bases. The 3000 base fragment is at double intensity to serve as a reference band. This ladder is suitable for use as an ssRNA size standard on denaturing or native agarose gels.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.