Learn More
Andwin Scientific SQUARE MOLD 22X22MM 288/CS
Supplier: Andwin Scientific 70182
The square Peel Away Disposable Embedding Molds are ideal for plastic or paraffin embedding. Block trimming is no longer required. This product has a dimension of 22 x 22 millimeters. It is sold in a case of 288.