Learn More
Andwin Scientific SPRING END BRUSHES 5/PK
Supplier: Andwin Scientific 17145020
The Flask and Bottle Brushes have white, natural bristles that are mounted on a flexible plastic handle. The handle's angled design allows users to each past narrow necks. This product is recommended for cleaning 250 mL volumetric flasks. The brush h