Learn More
MilliporeSigma™ Supelco™ SPME Fiber Assembly, Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm
For Gases and Low Molecular Weight Compounds (MW 30-225)
Supplier: MilliporeSigma™ Supelco™ FFA57295U
Description
Solid phase microextraction (SPME) technique is generally used for GC/MS analysis. It helps in simplifying and speeding up the sample preparation. It also reduces solvent requirement during extraction.Specifications
SPME | |
Solid Phase Micro-Extraction (SPME) Fibers & Accessories | |
23 ga |
Multi-Fiber Exchanger | |
Plain light blue hub | |
3/Pk. |