Learn More
Enzo Life Sciences Sphingomyelin (100 mg). CAS: 85187-10-6
SDP
Supplier:  Enzo Life Sciences BMLSL1350100
Encompass_Preferred
Contains primarily stearic and nervoic acids. Precursor to ceramide second messengers via the action of sphingomyelinase. Purity: ≥98% (TLC). Solubility: Soluble in chloroform, ethanol or methanol. Long Term Storage: -20°C.
Catalog No. 50-201-2115
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.