Learn More
Enzo Life Sciences Spermine NONOate (5mg). CAS: 136587-13-8
Supplier: Enzo Life Sciences ALX4300135005
Nitric oxide (NO) donor. A convenient reagent for preparing aqueous solutions of NO. Alternative name: SPER/NO, (Z)-1-N-[3-Aminopropyl]-N-[4-(3-aminopropylammonio)butyl]-amino-diazen-1-ium-1,2-diolate. Purity: ≥97% (NMR). Solubility: Soluble in water, methanol or 100% ethanol. Long Term Storage: -80°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.