Learn More
Electron Microscopy Sciences Speedy Foil, 2 x 2"
Supplier: Electron Microscopy Sciences 6040912
Pharmaceutical grade, pre-cut, interleaved aluminum foil squares. Time saving product that eliminates the wasteful practice of trimming pieces from over-sized rolls. Each size ships in its own ideally sized dispensing box that ensures squares remain crisp and flat. Squares are pre-cut to fit common laboratory container openings saving time. Ideal for test tubes, vials.
Size: 2 x 2"
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.