Learn More
Med Vet International Miltex MeisterHand Barraquer Eye Speculum, Large, 1.75in, German, MH18-35
Supplier: Med Vet International MILMH1835
Commonly used ophthalmologically when visualization of the globe is needed. Both end pieces of the blade can be fit into the upper and lower lid. Solid blades measuring out to 15mm. 1.75 in total. Large. MeisterHand build quality ensures superior craftsmanship. Made with high quality German steel.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.