Learn More
United Scientific™ Spectrum Tubes
SDPSCI_ED
The classic demonstration of spectral lines.
Supplier:  United Scientific™ SPTCL01
 View more versions of this product
Catalog No. S07001
Explore all special offers
Add to Cart
Description
Description
Acquaint students with the spectra of common gases in their pure form while demonstrating concept of wavelength.
  • 25 cm (10 in.) long
  • Capillary portion about 4 in. (10 cm) long
Specifications
Specifications
Chlorine Spectrum Tube
Emits characteristic spectrum when energized by spectroscope
Sealed electrode, metal endcap, loop and activity guide
25.4 cm
10.16 cm
5 to 12
Spectrometer
10 in.
4 in.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.