Learn More
MTC Bio Specimen/Transport Vial, 17x50mm, 7mL, PS, non-graduated, with attached screw-cap, non-sterile, CE, non-toxic, bulk pack of 700
Supplier: MTC Bio V2250
MTC’s 7mL Specimen Vials are supplied in translucent, virgin polypropylene or crystal clear virgin polystyrene. Both versions are supplied with an attached polyethylene screw cap. They are CE marked and certified leak proof. Packaged 700 vials with attached caps per case. They are mde of Virgin Polypropylene/Polystyrene, are non-cytotoxic, have a leakproof cap, and come in economical bulk packaging. RNase, DNase, pyrogen, & endotoxin free
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.