Learn More
Electron Microscopy Sciences EMS SEM Orbital Specimen Holder M4 Mount
Supplier:  Electron Microscopy Sciences 75912
Encompass_Preferred
The EMS Orbital Specimen Holder's clever design allows you to effortlessly rotate the specimen completely around an axis. The pot is coated to reduce noise, producing a black background behind the subject so every detail stands out, allowing the capture of stunning, clear images. The pot is detachable and fully repositionable below the pivoted mount. The position and hold of the sample can be controlled with the simple turn of a screw.
Catalog No. 50-192-7772
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.