Learn More
Andwin Scientific SPECIMEN CONT. 4.5OZ/S/BULK
SDP
Supplier:  Andwin Scientific 25384144
Encompass
The 133 mL specimen container is made from sturdy polypropylene that can provide secure storage for most liquid and solid specimens. The product is translucent and is compatible to keep strong acids, bases, and most organic solvents. It also features graduations in both voirdupois and metric units. This product is sold in a case of 100 containers with polyethylene purple caps measuring 57.15 mm in diameter.
Catalog No. NC9755953
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.