Learn More
Bel-Art™ SP Scienceware™ Pipette Pump™ Pipetter
Provides safe, accurate and trouble-free pipetting. Bel Art™ SP Scienceware™ Pipette Pump™ Pipetters utilize an easy-to-use one-handed operation.
Supplier: Bel-Art™ F378990000/EMD
Description
- Pipettes fit smoothly into its flexible tapered chuck
- Thumbwheel on side is rotated for precision aspirating or dispensing
- Plunger may be pressed for quick emptying
- Resistant to acids and alkalis
- Disassemble all parts for cleaning and before autoclaving at 121°C (250°F) for 20 min.; chuck should not be autoclaved
- Replacement Silicone Rubber Chuck and Washers are available
Specifications
Not Autoclavable | |
25 mL | |
Pipettes |
Pipette | |
Acid and Alkali | |
Red |