Learn More
STEMCELL Technologies Soybean Trypsin Inhibitor, ACF, Size: 1 g
Supplier: STEMCELL Technologies 07457
Soybean Trypsin Inhibitor, Animal Component-Free (ACF) is obtained from cultures free of animal-derived materials. This trypsin inhibitor consists of a single polypeptide chain with two disulfide bridges and inactivates trypsin on an equimolar basis. It is able to inhibit chymotrypsin to a lesser extent, but is ineffective against esterolytic, proteolytic, or elastolytic activities of porcine elastase (Vered et al.). Soybean Trypsin Inhibitor has also been shown to inhibit human and bovine thrombin, plasmin, and leukocytic proteases. Products are for Research Use Only, Not for Diagnostic or Therapeutic use, and are not to be administered to humans.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.