Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-10 Rat anti-Human, Viral, Cynomolgus, Unconjugated, Clone: JES3-12G8, Southern Biotech™
Rat Monoclonal Antibody
Supplier: Southern Biotechnology Associates 1011001
Description
SouthernBiotech cytokine antibodies offer high sensitivity and low background for use in a variety of applications.
Specifications
IL-10 | |
Monoclonal | |
0.5mg/mL | |
Supplied in borate buffered saline, pH 8.2; No preservatives or amine-containing buffer salts added | |
COS-expressed human IL-10 | |
RUO | |
Cynomolgus, Human, Virus | |
Liquid |
ELISA, Dot Blot, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
JES3-12G8 | |
Unconjugated | |
Rat | |
0.5 mg | |
Primary | |
Store at 2-8°C | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction