Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD8 Mouse anti-Human, LE/AF (Low Endotoxin/Azide Free), Clone: RFT8, Southern Biotech™
Mouse Monoclonal Antibody
Supplier: Southern Biotechnology Associates 953614
Description
SouthernBiotech anti-human primary antibodies are available in a variety of conjugated formats including Alexa Fluor and Pacific Blue.
Specifications
CD8 | |
Low Endotoxin/Azide-free (LE/AF) | |
0.5mg/mL | |
Mouse | |
0.5 mg | |
Primary | |
Store at -20°C |
Depletion Assay, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
RFT8 | |
Supplied in PBS | |
Thymocytes and E-rosetted lymphocytes | |
RUO | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction