Learn More
Cole-Parmer AEMC CA832 Sound Meter with Shockproof Holster
Supplier:  Cole-Parmer 8653320
Encompass
Lightweight meter features an ergonomically designed housing for comfortable, easy one-handed operation. Meter measures sound according to the sensitivity of the human ear with three dB measurement ranges, fast and slow responses, and both A and C weighting curves. Mount on tripod for stand-alone monitoring (tripod sold separately). What's included: microphone windscreen, tripod mounting screw, calibration tool, shockproof holster, and one 9 V battery. Compliance: ANSI S1.4-1983 Type 2, IEC 651 Type 2
Catalog No. 50-992-653
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.