Learn More
Revvity Health Sciences Inc Membrane Target Systems: Somatostatin sst5 (human) membrane preparation, in CHO-K1 cells. 400 Units.
Supplier:  Revvity Health Sciences Inc ES522M400UA
Encompass_Preferred
Membrane Target Systems are quality assured frozen membranes from cells that express recombinant or endogenous receptors. We submit every batch of receptor to stringent quality control testing that includes saturation radioligand binding assay to determine receptor concentration (Bmax) and affinity (Kd). Competition binding assays are performed determine affinity (Ki) against known reference agonists and antagonists. GTPgS data is also provided for some of our Gi coupled receptors. Membranes are carefully prepared and ready for a variety of HTS applications, including radioligand binding (using either proximity methods, such as FlashPlate, or classical filtration methods). Products are packaged as frozen crude membrane preparations. One assay unit is defined as micrograms of protein, defined by competition binding assay (filtration). A complete product description and recommended protocol are included on the Certificate of Analysis. Some of our receptors may be restricted for sale in
Catalog No. 50-904-9637
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.