Learn More
Hach Company Sodium Thiosulfate Standard Solution, Stabilized, 0.00246 N, 500 mL
Supplier: Hach Company 2408549
For determination of Chlorine by drop count titration. Used in Hach CN-65 Test Kit (225401). 500 mL bottle