Learn More
Quality Biological Inc Sodium Bicarbonate Solution, PK 4 X 100 mL
SDP
Supplier:  Quality Biological Inc 118085721
Encompass_Preferred
Sodium Bicarbonate (NaHCO3) is typically used in cell culture media as a buffer. ¿ Endotoxin and Mycoplasma Tested ¿ Although NaHCO3 does not have the buffering capacity of HEPES, it is occasionally preferred over HEPES because of its lower toxicity and nutritional value to cells in culture. ¿ Applications: Sodium Bicarbonate, 7.5% is useful as a buffer for culturing a broad spectrum of mammalian cells. ¿ Recommendation: dilute Sodium Bicarbonate in appropriate cell culture media, to a final concentration of 5-15 mM ¿ Storage Conditions: 15¿C to 30¿C ¿ Shipping condition: ambient temperature ¿ Sterile Filtered ¿ Endotoxin and Mycoplasma Tested ¿ Osmolality: 290 ¿ 5% mOsM/kg H2O ¿ Concentration: 0.893 mol/L ¿ Product and/or packaging contains recycled/renewable content ¿ Product and/or packaging is recyclable or reduces waste ¿ Product and/or packaging is safer to human and environmental health ¿ ¿ Max Shelf Life: 24 months
Catalog No. 50-983-212
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.