Learn More
*PMW INC B. Braun #L8002 Sodium Chloride, Preservative Free 0.9% IV Solution 250 ML SOD CHL, IVSOL PVC DEHP FREE 0.9% 250ML (24/CS)
Supplier:  *PMW INC 00264780020
Encompass_Preferred
Sodium Chloride Injections USP are sterile, nonpyrogenic, isotonic and contain no bacteriostatic or antimicrobial agents, PVC-Free and DEHP-Free container, Excel flexible plastic bag, These intravenous solutions are indicated for use in adults and pediatric patients as sources of electrolytes and water for hydration
Catalog No. NC3261467
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.