Learn More
Bioworld SOB Medium, 1 L
Supplier:  Bioworld 306210242
Encompass_Preferred
SOB MediumNutritionally rich growth medium for preparation and transformation of competent Escherichia coli cellspH: 7.0Dehydrated Appearance: light beige, homogeneous powderPrepared Appearance: light to medium amber brothSoluble in waterCan be used to prepare SOC Medium by adding 20 ml of 20% Dextrose Contains:Bacto Tryptone Bacto Yeast Extract Sodium Chloride Magnesium Sulfate Potassium Chloride
Catalog No. 50-254-447
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.