Learn More
MilliporeSigma™ Supelco™ Snyder Columns
Utilize separate chambers to promote multiple condensations and vaporizations during the distillation process, leading to a more efficient separation
Supplier:  MilliporeSigma™ Supelco™ 64721
 View more versions of this product
Catalog No. 11-100-4907
Description
Description
Specifications
Specifications
1 Ea.
Condensations and Vaporizations
Ground Joint
0-Ball Design
Snyder Column
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.