Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNRNP200 Polyclonal Antibody, Bethyl Laboratories
SDP

Rabbit Polyclonal Antibody

$150.28

Specifications

Antigen SNRNP200
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Liquid
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-156-5178
SDP
Encompass_Preferred
View Documents
Bethyl Laboratories, Inc
A303454AT
20 μL
Each of 1 for $150.28
Add to Cart
 
Description

Description

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predict

Pre-mRNA splicing is catalyzed by the spliceosome, a complex of specialized RNA and protein subunits that removes introns from a transcribed pre-mRNA segment. The spliceosome consists of small nuclear RNA proteins (snRNPs) U1, U2, U4, U5 and U6, together with approximately 80 conserved proteins. U5 snRNP contains nine specific proteins. This gene encodes one of the U5 snRNP-specific proteins. This protein belongs to the DEXH-box family of putative RNA helicases. It is a core component of U4/U6-U5 snRNPs and appears to catalyze an ATP-dependent unwinding of U4/U6 RNA duplices. Mutations in this gene cause autosomal-domit retinitis pigmentosa. Alternatively spliced transcript variants encoding different isoforms have been found, but the full-length nature of these variants has not been determined.
Specifications

Specifications

SNRNP200
Polyclonal
Liquid
RUO
TBS with 0.1% BSA and 0.09% sodium azide
activating signal cointegrator 1 complex subunit 3-like 1, ASCC3L1, bad response to refrigeration 2 homolog, BRR2, BRR2 homolog, HELIC2, KIAA0788, retinitis pigmentosa 33 (autosomal domit), RP33, small nuclear ribonucleoprotein 200kDa (U5), small nuclear ribonucleoprotein, U5 200kDa subunit, U5 snRNP specific protein, 200 KD, U5 snRNP-specific 200 kDa protein, U5-200KD
Between 400 and 450
Primary
4° C
SNRNP200
Immunohistochemistry, Western Blot
Unconjugated
Rabbit
Human, Mouse
O75643, Q6P4T2
23020, 320632
IgG
Antigen affinity chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.