Learn More
Andwin Scientific SNAPSTRIP 8X0.2ML ASSTD 120PK
Supplier: Andwin Scientific T30352
The SnapStrip is an attached 8-strip of tubes with each tube in the strip having its own individual attached dome cap. Another unique and innovative feature of SnapStrip tubes is the user's ability to create a strip of less than 8 tubes with caps by simply breaking the strip where desired. A strip of 2 or more tubes is easier and safer to use and manipulate than are single tubes with caps. Flexibility: create a 4-strip today, create a 6-strip tomorrow. The individual tubes may also be used with the 0.2 or 0.5 mL adapters. Packaged as 120 Strips of 8, case has 10 packs of 120 strips of 8. Ulta-thin wall for optimum heat transfer Certified free from DNase, RNase, and detectable DNA Assorted Color
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.