Learn More
New England Biolabs, Inc. SNAP-Cell TMR-STAR - 30 nmol
SDP
Supplier:  New England Biolabs, Inc. S9105S
Encompass
SNAP-Cell TMR-STAR - 30 nmol. SNAP-Cell TMR-Star is a red fluorescent substrate that can be used to label SNAP-tag fusion proteins inside living cells, on cell surfaces, or in vitro. This cell-permeable substrate (CP-6-TMR) is based on the single isomer 6-carboxytetramethylrhodamine and is suitable for standard rhodamine filter sets. It has an excitation maximum at 554 nm and an emission maximum at 580 nm. This package contains 30 nmol of SNAP-Cell TMR-Star substrate, sufficient to make 10 ml of a 3 M SNAP-tag fusion protein labeling solution.Help NEB go green. FREE FREIGHT on NEB orders greater than $350.00!
Catalog No. 50-811-2049
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.