Learn More
MTC Bio Snap Cap 5mL MacroTubes, non-sterile, four bags of 50 tubes (200 total). CLEAR
Supplier: MTC Bio C2500
This item has a minimum qty of 4 per supplier requirements.
MTC Bios new 5mL Five-O™ MacroTubes™ take all of the conveniences of standard 15mL conical tubes and scales them down for space efficiency and decreased natural resource consumption. Useful from -86C to 110C, these tubes are a valuable resource for boiling and for sample preservation at very low temperatures. Choose from gamma sterilized racked packaging with attached caps or non-sterile bulk packaging with separate caps. The foam racks are similar to the packaging normally associated with 15mL tubes (upright with caps attached), but they require about 60% less storage space. *Color: CLEAR. Tubes are not assorted colors*
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.