Learn More
World Precision Instrument Smallest Fiber Optic Dipping Probe-DIP-UV-MINI-5
SDP
Supplier:  World Precision Instrument DIPUVMINI5
Encompass_Preferred
Mini DipTip™s  outer diameter is only 1.5 mm—the size of a 17-gauge needle. It will fit into all micro centrifuge tubes on the market and is a useful tool for measuring protein and DNA samples. The diminutive dipping probe can also be used for a dissolution system.
Catalog No. 50-253-8869
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.