Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ Safety-Walk™ Slip Resistant General Purpose Tape, Series 600
Mineral coated, slip resistant material features a highly durable surface. Low profile design helps reduce tripping hazards.
Supplier: 3M™ Safety-Walk™ 7000052247
This item is not returnable.
View return policy
Description
- Anti-slip tape for light to heavy shoe-traffic areas
- High quality abrasive particles to ensure long-lasting friction in any application
- Highly durable surface withstands the rigors of equipment traffic
- High performance, pressure-sensitive adhesive for easy application
- Custom shapes and sizes are also available
- NFSI High Traction Certification
Specifications
Mineral-coated | |
Light to Heavy Traffic Areas | |
4 Pk. | |
25 mm | |
Yellow |
18.3 m x 25 mm | |
18.3 m | |
1 in. | |
Pressure Sensitive Adhesive | |
Floor Tape |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Share a Content Correction