Learn More
Newcomer Supply StainTray 10 Place with Clear Lid. Holds up to 10 slides on plastic rails covered with a polymer strip to hold slides even at an angle.
SDP
Supplier:  Newcomer Supply 684810CL
Encompass
The Simport StainTray™ has a black base made of tough ABS plastic withstanding a wide range of chemicals (Avoid chlorinated hydrocarbons ). It will accept up to 10 slides on four plastic rails covered with a red polymer strip to perfectly hold slides even if tray is held at an angle. When humidity is needed, wells between rails will hold up to one ml of water securely without splashing. Middle wells will hold up to 2 ml each. Rails are raised not only to avoid water touching the slides but to make them more easily retrieveable. The base will also hold excess stain solution dripping from the slides. Four rubber feet ensure greater base stability. The lid is Clear allowing for visual examination and is made of PETG with a temp. range of -20°C to +60°C. Units are stackable for space saving purposes. Not autoclavable. Dimensions: 24 x 24 x 4.5 cm (H).
Catalog No. NC2102251
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.