Learn More
Electron Microscopy Sciences Plastic 2-Place Slide Mailer Assorted
Supplier:  Electron Microscopy Sciences 7155601
Encompass_Preferred
These disposable slide mailers are ideal for secure shipping and storing of slides. Made from polypropylene, the protectors hold two standard slides (25 x 75 mm and 1 x 3"), with large snaps that are easy to secure.
Catalog No. 50-192-9415
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.