Learn More
Electron Microscopy Sciences Globe Slides Two-Sided Frosted, Ground Edge, 90° Corner
Supplier:  Electron Microscopy Sciences 7188325
Encompass_Preferred
These frosted microscope slides are produced using a unique chemical etching process. This provides the slides with a smooth frosted finish that won't wear out slide marking pens.
Edge: Ground
Corner: 90°
Catalog No. 50-192-8792
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.