Learn More
Electron Microscopy Sciences Globe Microscope Slides, Beveled Edge, Clipped Corner, Yellow Frosted Label
Supplier: Electron Microscopy Sciences 7188375
Color frosted microscope slides with color coding on one end and one side. Pre-cleaned and ready to use. Edges specially angled at 45 degrees to provide secure grasp during blood smearing process.
Edge: Beveled
Corner: Clipped
Label: Yellow Frosted