Learn More
Electron Microscopy Sciences Globe Plain Slides, Beveled Edge, Clipped Corner
Supplier:  Electron Microscopy Sciences 7188317
Encompass_Preferred
These plain microscope slides are produced from premium glass sheets and offer exceptional quality while providing considerable savings. These slides are pre-cleaned and ready to use. Dimensions: 25 x 75 mm. Thickness: 1.1 mm
Edge: Beveled
Corner: Clipped
Catalog No. 50-192-8784
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.