Learn More
Andwin Scientific SLIDE BASKET 10/CS
Supplier: Andwin Scientific 4768
The Prisma 20-Slide Basket is an accessory for Tissue-Tek Prisma, Film and G2 Coverslippers, a ready-to-use product that offers superior performance, convenience, and flexibility in staining and glass coverslipping. This product by Sakura features a quick action of coverslipping 360 slides per hour. It is sold separately. The slide basket is sold in a case of 10.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.