Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC7A5 Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 286701AP150UL
Description
SLC7A5 Polyclonal antibody specifically detects SLC7A5 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
SLC7A5 | |
Polyclonal | |
Unconjugated | |
Q01650 | |
Rabbit | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
0.9 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
4F2LC; CD98; CD98LC; D16S469E; E16; hLAT1; LAT1; MPE16; SLC7A5 | |
Antigen affinity chromatography | |
RUO | |
8140 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction