Learn More
Andwin Scientific SINGLE EDGE RAZOR BLADE 100/PK
SDP
Supplier:  Andwin Scientific 55411050
Encompass
The single-edge industrial razor blades are machine-honed from hard carbon steel. The blades have thickness of 0.22 mm (0.009"). This product is sold in a pack of 100 individually wrapped blades.
Catalog No. NC9148528
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.