Learn More
Electron Microscopy Sciences Silicone Staining Pad 5" x 3" x ½"
Supplier: Electron Microscopy Sciences 71565
Made from white silicone, a non-reactive material. Pad has 40 cells in 5 rows of 8 each. Each cell is half-sphere shaped with an opening of 6 mm dia. and 5 mm deep. A few drops of staining solution is added to the wells and grids are then immersed and retrieved as per staining procedure. A watch-glass plate comes with each dish to reduce oxygen and evaporation.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.