Learn More
Electron Microscopy Sciences Cold Embedding Silicone Mold for Acylic Resin 38 mm
Supplier: Electron Microscopy Sciences 5101238
This mold is for acrylic resins only and the best mounting cup for all of our acrylic resins. The flexible silicone rubber makes it very easy to remove the mount from the mounting cup.
Diameter: 38 mm