Learn More
Electron Microscopy Sciences Silica Gel Packets 3 Gram, 1-1/16" x 2-1/16", 24.4 cubic inch Capacity
Supplier:  Electron Microscopy Sciences 7120603
Encompass_Preferred
Silica Gel-Desiccant is a fast and extra moisture absorbing agent. Meets FDA specifications for food and drug product. Provides superior absorption. Remains dry at maximum saturation. Silica gel can absorb 15% of its weight in water vapor in two hours.
Dimensions: 1-1/16" x 2-1/16"
Capacity: 24.4 cubic inch
Catalog No. 50-930-890
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.