Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Public Seating™ Signature Science Lab Table, Light Oak Legs
SCI_ED

NPS Signature Frame Adjustable Height Science Lab Tables have perfect chemistry with any science classroom.

Supplier:  National Public Seating™ SLT52460C

 View more versions of this product

Catalog No. S83703


Explore all special offers
Add to Cart

Description

Description

  • Round Legs have (2) 3/8 in. Diameter in Textured Black
  • Legs Adjust Smoothly and Easily in 3/4 in. Increments – from 27 to 42 in. Height
  • Adapts for ADA Requirements
  • Easy Assembly, With Pre-Drilled Pilot Holes
  • Patented Mechanism Allows Table to Rise Effortlessly and Automatically Lock At Your Height Setting Requirement
  • Weight Capacity: 500 lb.
  • Oversized Matching Floor Glides are Standard
  • 10 Year Warranty
Specifications

Specifications

Lab Table
Black
Science Classrooms
10 years
500 lb.
MAS2, ANSI BIFMA
Resistant to water and 13 common lab chemicals
Chemical Resistant
60 x 24 x 27 to 42 in.
Oak woodgrain legs
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.