Learn More
Enzo Life Sciences Shrimp alkaline phosphatase, (recombinant) (5000U)
Supplier: Enzo Life Sciences ENZPRT1615000
Shrimp alkaline phosphatase (recombinant), (rSAP) is a multipurpose alkaline phosphatase that is fully inactivated by a short heat treatment. rSAP has all the properties of the native enzyme, but with the additional benefits of greater stability, consistency, and purity. rSAP simplifies cloning and sequencing workflows due to a complete heat inactivation. Shrimp alkaline phosphatase (recombinant), (rSAP) is a multipurpose alkaline phosphatase that is fully inactivated by a short heat treatment. rSAP has all the properties of the native enzyme, but with the additional benefits of greater stability, consistency, and purity. rSAP simplifies cloning and sequencing workflows due to a complete heat inactivation. Alternative name: rSAP. Concentration: 1U/µl for 1000 unit size 5U/µl for 5000 unit size. Formulation: Liquid. Source: Pandalus borealis (Northern red shrimp) origin, recombinantly produced in Pichia pastoris. Long Term Storage: -20°C. Applications: Cloning.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.