Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NOTRAX™ Shoe Sanitizer System
These shoe sanitizing systems provide an effective solution to minimize the spread of contaminants tracked on the bottoms of shoes.
Supplier: NOTRAX™ 355S002700
Description
- Simple, safe and effective shoe disinfecting system to limit the spread of contaminants on floors
- Labels clearly instruct how to sanitize shoes
- Ideal for entrances to schools, businesses, facilities, and more
- Low profile bevels for easy walk on/off to minimize trip hazards
- Designed as a zoned system with completely contained and secured components
- Carpet mat inserts are disposable and replaceable for quick and easy cleaning
- Available in 2-zone (disinfecting, drying) and 3-zone (cleaning, disinfecting, drying) options
- Each system cleans and sanitizes shoe soles, then quickly dries them the moment you step out
- Mat inserts are adhesive backed and replaceable for easy cleaning
- Carpet mats are made from recycled plastic bottles
- System is lightweight and easy to set up and move
- Vinyl backed to prevent slipping
Specifications
27 in. | |
108.5 x 27 in. | |
68.6 cm | |
Black | |
Recycled Plastic |
108.5 in. | |
275.6 cm | |
Entrance Mat | |
Shoe Cleaning, Disinfecting, and Drying System (3-Zone) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction