Learn More
Showa Denko America Inc SHODEX OHPAK SB-807 HQ COLUMN
Supplier:  Showa Denko America Inc F6429108
Encompass_Preferred
Polyhydroxymethacrylate columns for aqueous SEC (GFC) analysis. The eluent can be replaced with DMF for the analysis of polar polymers. Good replacement for protein analysis when silica cannot be used. Pullulan exclusion limit: 500,000,000
Catalog No. NC9045703
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.