Learn More
Rockland Immunochemicals Sheep Blood In Heparin Sodium 100 mL
Supplier: Rockland Immunochemicals R5110100
Sheep whole blood is aseptically collected and prepared to order from our colony of animals. These cell suspensions are useful for the titration of complement adsorption procedures testing for agglutinins/HA assays and for the preparation of stroma as particulate reagents. Sheep red blood cells are perishable and are collected and processed upon receipt of your order.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.