Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SGE™ 2.5mL Syringe with removable Luer Lock valve and GT plunger

Prefitted with Luer Lock dispenser valve.

Supplier:  SGE™ 008560

Catalog No. SG008560


Description

Description

 

  • Low syringe carryover
  • Reduced nonspecific sample interaction
  • Increased analyte recovery
  • Ideal for sample storage and transportation or to pressurize sample prior to injection
  • Material: Borosilicate glass, PTFE plunger (Gas tight)
Note: Needle is not included; sold separately.
Specifications

Specifications

Autoclavable
For sample storage and transportation or to pressurize sample prior to injection
2.5 mL
Borosilicate Glass
Syringe
Luer Lock, Abnehmbar
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.